Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUS10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$464.00
Specifications
Antigen | PUS10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
PUS10 Polyclonal specifically detects PUS10 in Human samples. It is validated for Western Blot.Specifications
PUS10 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
CCDC139, coiled-coil domain containing 139, Coiled-coil domain-containing protein 139, DOBIMGC126729, EC 5.4.99, EC 5.4.99.-, FLJ32312, MGC126755, pseudouridine synthase 10, pseudouridylate synthase 10, Psi55 synthase, PUS1, putative tRNA pseudouridine synthase Pus10, tRNA pseudouridine 55 synthase, tRNA pseudouridylate synthase, tRNA-uridine isomerase | |
PUS10 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q3MIT2 | |
150962 | |
Synthetic peptides corresponding to PUS10 (pseudouridylate synthase 10) The peptide sequence was selected from the middle region of PUS10. Peptide sequence AVFVAGRYNKYSRNLPQTPWIIDGERKLESSVEELISDHLLAVFKAESFN. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title