Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ PVC Motor Blower
SDPSCI_ED

Labconco™ PVC Blower provide optimum chemical resistance and are made specifically to handle high order oxidizers such as perchloric acid.

Supplier:  Labconco™ 7183100/EMD

 View more versions of this product

Catalog No. S28340

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View alternate product


Description

Description

  • Housings are vacuum formed from unplasticized Type I polyvinyl chloride for complete corrosion protection
  • Flow-through vent holes and louvers allow for air circulation that cools motor, thus prolonging motor life
  • Impellers are made of injection molded PVDF to resist highly corrosive atmospheres
  • Powder-coated base and weather cover provide long life in extreme weather
  • Inlet and discharge diameters are sized to receive rigid PVC ducting material directly for easy installation
  • Blowers are available with 25.4cm (10 in.) and 30.4cm (12 in.) inlet/outlet connections
  • Sheaves are adjustable to save energy and to allow for air volume adjustment
  • Engineered for quiet operation
  • Sound levels generally range between 64 and 88 decibels depending on blower size
  • Intelli-Sense models range from 50 to 70dB
  • Impeller is bonded directly to shaft to reduce maintenance and increase life
  • Self-adjusting gravity belt tightener insures consistent performance because belt is held taut by weight of motor
  • Motor mount keeps motor in place
  • Convenient access to impeller assembly provides for easy maintenance
  • Variable exhaust orientation adjusts to three outlet positions for installation flexibility. Intelli-Sense models allow eight positions (vertical discharge is recommended)
Specifications

Specifications

90 lb.
0.5 hp
115 V, 60 Hz, Single Phase, 8.4 A
881 to 1248 rpm
Belt-driven
41 Kg
Outlet: 27.3 cm (10.75 in.); Inlet: 26.3 cm (10.38 in.)
PVC (Type I)
Perchloric Acid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit