Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PVR (CD155) Mouse anti-Human, Clone: L95, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABF2734100UG
Description
PVR (CD155) Monoclonal antibody specifically detects PVR (CD155) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition, Neutralizing.Specifications
PVR (CD155) | |
Monoclonal | |
1.0 mg/mL | |
Purified mouse monoclonal antibody IgG1 in PBS without azide | |
Human embryonic 293T cells. | |
100 μg | |
Primary | |
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Inhibition Assays, Neutralization | |
L95 | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction