Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVR (CD155) Mouse anti-Human, Unconjugated, Clone: TX24, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen PVR (CD155)
Applications Flow Cytometry
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF23381MI
View Documents
MilliporeSigma
MABF2338100UG
100 μg
Each for $372.00
Add to cart
 
MABF23382MI
View Documents
MilliporeSigma
MABF233825UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

PVR (CD155) Monoclonal antibody specifically detects PVR (CD155) in Human samples. It is validated for Flow Cytometry, Inhibits Activity/Function.
Specifications

Specifications

PVR (CD155)
Unconjugated
Mouse
Poliovirus receptor;Nectin-like protein 5;NECL-5;CD155
A recombinant fragment corresponding to 323 amino acids from the extracellular domain of human Poliovirus receptor (CD155).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Flow Cytometry
Purified
Human
PVR, PVS
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit