Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVR (CD155) Rat anti-Mouse, Unconjugated, Clone: TX56, MilliporeSigma™

Rat Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen PVR (CD155)
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF23491MI
View Documents
MilliporeSigma
MABF2349100UL
100 μL
Each for $372.00
Add to cart
 
MABF23492MI
View Documents
MilliporeSigma
MABF234925UL
25 μL
Each for $147.00
Add to cart
 
Description

Description

PVR (CD155) Monoclonal antibody specifically detects PVR (CD155) in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation, Inhibits Activity/Function, Western Blotting.
Specifications

Specifications

PVR (CD155)
Unconjugated
Rat
Poliovirus receptor
CD155 transfected cells.
Primary
Stable for 1 year at -20°C from date of receipt.
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Purified
Mouse
Pvr, cd155, D7ertd458e
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit