Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PvRBP2b Mouse anti-Plasmodium vivax, Unconjugated, Clone: 6H1, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen PvRBP2b
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF19941MI
View Documents
MilliporeSigma
MABF1994100UL
100 μL
Each for $386.00
Add to cart
 
MABF19942MI
View Documents
MilliporeSigma
MABF199425UL
25 μL
Each for $153.50
Add to cart
 
Description

Description

PvRBP2b Monoclonal antibody specifically detects PvRBP2b in Plasmodium vivax samples. It is validated for ELISA, Flow Cytometry, Inhibits Activity/Function, Immunoprecipitation, Western Blotting.
Specifications

Specifications

PvRBP2b
Unconjugated
Mouse
Reticulocyte-binding protein 2
His-tagged recombinant fragment corresponding to 1294 amino acids from the N-terminal half of Plasmodium vivax Reticulocyte-binding protein 2 (PvRBP2b).
Primary
Stable for 1 year at -20°C from date of receipt.
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Purified
Plasmodium
RBP-2, RBP2
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit