Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVRIG Rabbit anti-Human, Alexa Fluor™ 488, Clone: 2334A, R&D Systems

Rabbit Monoclonal Antibody

Manufacturer:  R&D Systems FAB93651G100UG

Catalog No. FAB93651G10


Add to cart
This item is not returnable. View return policy

Description

Description

PVRIG Monoclonal antibody specifically detects PVRIG in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

PVRIG
2334A
Flow Cytometry 0.25-1 μg/106 cells
Q6DK17
C7orf15, C7orf15MGC138295, CD112R, MGC104322, MGC138297, MGC2463, poliovirus receptor related immunoglobulin domain containing, Poliovirus receptor-related immunoglobulin domain-containing protein, transmembrane protein PVRIG
Human embryonic kidney cell, HEK293-derived human PVRIG protein, Thr 41 & Glu43-Leu172, Accession No. Q6DK17
Protein A or G purified from cell culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects human PVRIG in direct ELISAs.
Flow Cytometry
Alexa Fluor 488
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Conjugated, Purified, Supernatant
Rabbit
IgG
100 μg
Primary
79037.0
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit