Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PVRIG, Rabbit anti-Human, Clone: 2334B, R&D Systems™

Rabbit Monoclonal Antibody

$131.70 - $315.00

Specifications

Antigen PVRIG
Clone 2334B
Applications Flow Cytometry, Blocking Assay, CyTOF
Host Species Rabbit
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB93652100
View Documents
R&D Systems
MAB93652100
100 ug
Each for $315.00
Add to cart
 
MAB93652SP
View Documents
R&D Systems
MAB93652SP
25 ug
Each for $131.70
Add to cart
 
Description

Description

PVRIG Monoclonal antibody specifically detects PVRIG in Human Samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF.
Specifications

Specifications

PVRIG
Flow Cytometry, Blocking Assay, CyTOF
Human
Primary
2334B
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit