Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human PVRL4 Partial ORF (NP_112178, 31 a.a. - 139 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Manufacturer: Abnova™ H00081607Q01S
Description
Poliovirus receptor-like proteins (PVRLs), such as PVRL4, are adhesion receptors of the immunoglobulin superfamily and function in cell-cell adhesion (Reymond et al., 2001 [PubMed 11544254]).[supplied by OMIM]
Sequence: AGELGTSDVVTVVLGQDAKLPCFYRGDSGEQVGQVAWARVDAGEGAQELALLHSKYGLHVSPAYEGRVEQPPPPRNPLDGSVLLRNAVQADEGEYECRVSTFPAGSFQASpecifications
NP_112178 | |
wheat germ expression system | |
Liquid | |
LNIR|PRR4|nectin-4 | |
PVRL4 | |
AGELGTSDVVTVVLGQDAKLPCFYRGDSGEQVGQVAWARVDAGEGAQELALLHSKYGLHVSPAYEGRVEQPPPPRNPLDGSVLLRNAVQADEGEYECRVSTFPAGSFQA | |
PVRL4 (Human) Recombinant Protein (Q01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Yes | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
PVRL4 | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81607 | |
Wheat Germ (in vitro) | |
37.73kDa | |
GST | |
10 ug | |
RUO |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title