Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PVRL4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 219031AP150UL
Description
Poliovirus receptor-like proteins (PVRLs), such as PVRL4, are adhesion receptors of the immunoglobulin superfamily and function in cell-cell adhesion (Reymond et al., 2001 ).Specifications
PVRL4 | |
Polyclonal | |
Unconjugated | |
NECTIN4 | |
Ig superfamily receptor LNIR, LNIR, nectin 4, poliovirus receptor related 4, PRR4, PVRL4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
71740, 81607 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R007, Q96NY8 | |
Nectin4 | |
PVRL4 Fusion Protein Ag14127 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction