Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PWWP2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155176
Description
PWWP2B Polyclonal specifically detects PWWP2B in Human samples. It is validated for Western Blot.Specifications
PWWP2B | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
PWWP2B | |
Synthetic peptides corresponding to PWWP2B(PWWP domain containing 2B) The peptide sequence was selected from the N terminal of PWWP2B. Peptide sequence ILLDCTKKSGLFGLPPLAPLPQVDESPVNDSHGRAPEEGDAEVMQLGSSS. | |
Affinity purified | |
RUO | |
170394 | |
Human, Mouse, Rat, Pig, Bovine | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
bA432J24.1, FLJ39621, FLJ46823, pp8607, PWWP domain containing 2, PWWP domain containing 2B, PWWP domain-containing protein 2B, PWWP2, RP11-273H7.1 | |
Rabbit | |
53 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction