PXK Recombinant Protein, Abnova

Manufacturer: Abnova Corporation H00054899P0125

 View more versions of this product

Catalog No. 89-013-029

  Check Availability
Add to Cart

    Specifications

    Molecular Weight 75.24
    Name Human PXK Full-length ORF Recombinant Protein with GST-tag at N-terminal
    Purification Method Glutathione Sepharose 4 Fast Flow
    For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
    Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
    Quality Control Testing 12.5% SDS-PAGE stained with Coomassie Blue
    Quantity 25μg
    Recombinant Yes
    Source Wheat Germ (in vitro)
    Storage Requirements -80°C
    Cross Reactivity Human
    pH Range 8
    Preparation Method In vitro wheat germ expression system
    Format Solution

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit