Learn More
PXMP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 221631AP
Catalog No. 50-173-3752
Add to cart
Description
Description
This gene encodes an integral peroxisomal membrane protein required for peroxisome biogenesis. The protein is thought to be involved in peroxisomal matrix protein import. Mutations in this gene result in one form of Zellweger syndrome and infantile Refsum disease. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
Specifications
PXMP3
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P28328, P55098
PAF 1, PAF1, Peroxin 2, Peroxisomal membrane protein 3, Peroxisome assembly factor 1, Peroxisome biogenesis factor 2, PEX2, PMP3, PMP35, PXMP3, RING finger protein 72, RNF72
Rabbit
IgG
150 μL
-20°C
Primary
19302, 5828
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PEX2
Liquid
PEX2
PXMP3 Fusion Protein Ag17815
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse