Learn More
PXMP4 Rat anti-Mouse, Alexa Fluor™ 350, Clone: 7S41, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221987AF350
Catalog No. NP221987A30
Add to cart
Description
Description
PXMP4 Monoclonal antibody specifically detects PXMP4 in Mouse samples. It is validated for ELISA.
Specifications
Specifications
PXMP4
7S41
ELISA
PMP24, PXMP4 peroxisomal membrane protein 4, 24kDa
Rat
IgG2
Store at 4°C in the dark.
Monoclonal
Mouse
ELISA
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
PXMP4
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with mouse recombinant protein MMP9. Rat monoclonal anti mouse PXMP4 P21245.
0.1 mL
Primary
11264