Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PXR/NR1I2 Mouse anti-Human, Clone: 6H11D8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP261817

Catalog No. NBP2-61817



Description

Description

PXR/NR1I2 Monoclonal antibody specifically detects PXR/NR1I2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Specifications

Specifications

PXR/NR1I2
6H11D8
Unconjugated
Purified
nuclear receptor subfamily 1 group I member 2, nuclear receptor subfamily 1, group I, member 2, ONR1, Orphan nuclear receptor PAR1, Orphan nuclear receptor PXR, PAR, PAR1, PAR2, Pregnane X receptor, PRR, PXRPARq, SAR, Steroid and xenobiotic receptor, SXRBXR
NR1I2
Purified recombinant fragment of human NR1I2 (AA:1-142) expressed in E. Coli.
9.8 kDa
Primary
0.1 ml
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot,Flow Cytometry,ELISA
1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
PBS with 0.05% Sodium Azide
8856
Mouse
IgG1
Monoclonal
Protein G purified
Stem Cell Markers, Transcription Factors and Regulators
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit