Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PYCARD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA588132
Description
Immunogen sequence: LDLTDKLVSF YLETYGAELT ANVLRDMGLQ EMAGQLQAAT HQGSGAAPAG IQAPPQSAAK PGLHFIDQHR AALIARVTNV EWLLDALYGK VLTDEQYQAV RAEPTNPSKM RKLFSFTPAW NWTCKDLLLQ ALRESQSYLV EDLERS; Positive Samples: A-549, THP-1; Cellular Location: Cytoplasm, Endoplasmic reticulum, Mitochondrion, Nucleus
This gene encodes an adaptor protein that is composed of two protein-protein interaction domains: a N-terminal PYRIN-PAAD-DAPIN domain (PYD) and a C-terminal caspase-recruitment domain (CARD). The PYD and CARD domains are members of the six-helix bundle death domain-fold superfamily that mediates assembly of large signaling complexes in the inflammatory and apoptotic signaling pathways via the activation of caspase. In normal cells, this protein is localized to the cytoplasm; however, in cells undergoing apoptosis, it forms ball-like aggregates near the nuclear periphery. Two transcript variants encoding different isoforms have been found for this gene.Specifications
PYCARD | |
Polyclonal | |
Unconjugated | |
PYCARD | |
Pycard | |
Recombit fusion protein containing a sequence corresponding to amino acids 50-195 of human PYCARD | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry, Western Blot | |
1.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9EPB4, Q9ULZ3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
282817, 29108, 66824 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title