Learn More
PYCR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 221501AP
Catalog No. 50-173-3745
Add to cart
Description
Description
This gene encodes an enzyme that catalyzes the NAD(P)H-dependent conversion of pyrroline-5-carboxylate to proline. This enzyme may also play a physiologic role in the generation of NADP(+) in some cell types. The protein forms a homopolymer and localizes to the mitochondrion. Alternate splicing results in two transcript variants encoding different isoforms.
Specifications
Specifications
PYCR1
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P32322, Q922W5
P5C, P5C reductase 1, P5CR, P5CR 1, PIG45, PP222, PYCR, PYCR1
Rabbit
IgG
150 μL
-20°C
Primary
209027, 287877, 5831
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
PYCR1
Liquid
Pycr1
PYCR1 Fusion Protein Ag17745
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat