Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PYCR2 Mouse anti-Human, Clone: OTI4A10, liquid, TrueMAB™
Mouse Monoclonal Antibody
Supplier: OriGene TA501877S
Description
PYCR2 belongs to the pyrroline-5-carboxylate reductase family. The function of the PYCR2 protein is not known.Specifications
PYCR2 | |
Monoclonal | |
0.67 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96C36 | |
PYCR2 | |
Full length human recombit protein of human PYCR2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4A10 | |
Unconjugated | |
PYCR2 | |
HLD10, P5CR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29920 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction