Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PYCR2 Monoclonal Antibody (OTI2A5), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA525281
Description
PYCR2 Monoclonal antibody specifically detects PYCR2 in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
PYCR2 | |
Monoclonal | |
0.72 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96C36 | |
Pycr2 | |
Full length human recombinant protein of PYCR2 produced in HEK293T cell | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Western Blot | |
OTI2A5 | |
Unconjugated | |
Pycr2 | |
1810018M05Rik; HLD10; P5C reductase 2; P5CR 2; P5CR2; Pycr2; pyrroline 5-carboxylate reductase isoform; pyrroline-5-carboxylate reductase 2; pyrroline-5-carboxylate reductase family member 2; pyrroline-5-carboxylate reductase family, member 2; QtsA-16987; unnamed protein product | |
Mouse | |
Affinity Chromatography | |
RUO | |
29920 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction