Learn More
PYCRL Mouse anti-Human, Clone: OTI1C11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF502034

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PYCRL encodes a protein that belongs to the pyrroline-5-carboxylate reductase family of enzymes. Members of this family catalyze the final step in proline biosynthesis, converting pyrroline-5-carboxylate to proline. Glutamate and ornithine are precursors in the synthesis of proline. The protein encoded by this gene is a cytoplasmic enzyme involved in the biosynthesis of proline from ornithine.Specifications
PYCRL | |
Monoclonal | |
Unconjugated | |
PYCR3 | |
PYCRL | |
Mouse | |
Affinity Chromatography | |
RUO | |
65263 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1C11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q53H96 | |
PYCR3 | |
Full length human recombit protein of human PYCRL produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.