Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYCRL Mouse anti-Human, Clone: OTI3E1, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA502033

Catalog No. 50-167-3120


Add to cart

Description

Description

PYCRL encodes a protein that belongs to the pyrroline-5-carboxylate reductase family of enzymes. Members of this family catalyze the final step in proline biosynthesis, converting pyrroline-5-carboxylate to proline. Glutamate and ornithine are precursors in the synthesis of proline. The protein encoded by this gene is a cytoplasmic enzyme involved in the biosynthesis of proline from ornithine.
Specifications

Specifications

PYCRL
Monoclonal
0.81 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q53H96
PYCRL
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
65263
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI3E1
Unconjugated
PYCR3
Liquid
PYCR3
Full length human recombit protein of human PYCRL produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit