Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYGM Mouse anti-Human, Clone: OTI3F9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527445

Catalog No. PIMA527445


Add to cart

Description

Description

This gene encodes a muscle enzyme involved in glycogenolysis. Highly similar enzymes encoded by different genes are found in liver and brain. Mutations in this gene are associated with McArdle disease (myophosphorylase deficiency), a glycogen storage disease of muscle. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

PYGM
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P11217
PYGM, GPMM, Myophosphorylase, Phosphorylase, glycogen muscle
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5837
Immunohistochemistry (Paraffin), Western Blot
OTI3F9
Unconjugated
PYGM
Liquid
PYGM
Human recombit protein fragment corresponding to amino acids 698-842 of PYGM produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit