Learn More
Aviva Systems Biology Corp PYHIN1 Antibody - N-terminal region (ARP50875_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50875_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PYHIN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PYHIN1. Alternate names: IFIX, MGC23885, RP11-520H16.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60458