Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PyOxim, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
153433-21-7
C17H29N5O3P·F6P
527.39
RDWDVLFMPFUBDV-PXMDEAMVSA-N
pyoxim,pyoxime,ethyl cyano hydroxyimino acetato-tri-1-pyrrolidinyl-phosphonium hexafluorophosphate,cyano-ethoxycarbonyl methyleneaminooxy tris 1-pyrrolidinyl phosphonium hexafluorophosphate
46910530
[(E)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-tripyrrolidin-1-ylphosphanium;hexafluorophosphate
CCOC(=O)C(=NO[P+](N1CCCC1)(N2CCCC2)N3CCCC3)C#N.F[P-](F)(F)(F)(F)F
153433-21-7
C17H29N5O3P·F6P
527.39
RDWDVLFMPFUBDV-PXMDEAMVSA-N
pyoxim,pyoxime,ethyl cyano hydroxyimino acetato-tri-1-pyrrolidinyl-phosphonium hexafluorophosphate,cyano-ethoxycarbonyl methyleneaminooxy tris 1-pyrrolidinyl phosphonium hexafluorophosphate
46910530
[(E)-(1-cyano-2-ethoxy-2-oxoethylidene)amino]oxy-tripyrrolidin-1-ylphosphanium;hexafluorophosphate
CCOC(=O)C(=NO[P+](N1CCCC1)(N2CCCC2)N3CCCC3)C#N.F[P-](F)(F)(F)(F)F
Packaging Glass Bottle
Color White
Water 1% max.
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 96% min. (HPLC)
Formula Weight 527.39
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material PyOxim

GHS H Statement
Risk of explosion if heated under confinement.

GHS P Statement
Do not get in eyes,on skin,or on clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.