Learn More
Pyramex™ Ridgeline Ratchet Headgear
SDP
Ridgeline Headgear with ratchet and pivoting action
Manufacturer:  Pyramex™ HGB
Catalog No. 19-817-310
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
This item is not returnable. View return policy
Description
Description
  • Constructed from durable nylon
  • Universal pegs will accommodate a wide variety of face shields
  • Ratchet style adjustment for easy fit
  • Pivoting action allows user to lift face shield while wearing
  • Crown width: 6.25 in.
Specifications
Specifications
Black
Nylon