Learn More
Pyramex™ Rendezvous™ Safety Glasses
Adjustable lens angle aligns to the proper position for various work applications
Manufacturer:  Pyramex™ SCB2880S
 View more versions of this product
Catalog No. 19-121-012
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Ratcheting lens pitch adjustment aligns lens angle to properly position for work application.
  • Flexible nylon temples with four size settings to accommodate a broad range of facial sizes
  • Adjustable nose pad conforms to a broad range of bridge sizes
  • Lightweight nylon frame for comfortable, all day wear
  • Replaceable, 9.5 base-curved polycarbonate lens withstands high-impact
  • Provide 99% UV protection, is completely distortion free
Compliance
Meet ANSI Z87.1 requirements.
Specifications
Specifications
Meet ANSI Z87.1 requirements.
Polycarbonate
Indoor/Outdoor Mirror
Polycarbonate
Blue
Scratch-resistant