Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Lanyard
SDP

Catalog Number 19151638
Click to view available options
Product Type:
Breakaway Retainer
Clip-on Retaining Cord
Cotton Retaining Cord
Retaining Cord w/ Plastic Clips
Retaining Cord w/ Rubber Tips

For use in retaining spectacles

Fits a variety of temples.
Color Black
Product Type Retaining Cord w/ Plastic Clips
For Use With (Application) Retaining spectacles
Includes Plastic clips
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit