Pyramex™ V2 Readers Safety Glasses
Manufacturer: Pyramex™ SB1880R20
 View more versions of this product
Catalog No. 19-130-3484
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Indoor/outdoor application
    Magnification Power 2
    Surface Coating Scratch resistant
    Frame Color Black
    Lens Tint Mirror
    Material Polycarbonate
    UV Protection 99.90%
    Type Reader Spectacles
  • Description & Specifications
Related Products
Product Content Feedback