Pyramex™ V2 Readers Safety Glasses
Manufacturer: Pyramex™ SB1810R15
 View more versions of this product
Catalog No. 19-121-922
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Indoor application
    Frame Color Black
    Lens Tint Clear
    Magnification Power 1.5
    Material Polycarbonate
    Surface Coating Scratch resistant
    UV Protection 99.90%
    Type Reader Spectacles
  • Description & Specifications
Related Products
Product Content Feedback