Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrantel Pamoate, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
20 kg
Packaging:
Amber Glass Bottle
Fiber Drum
22204-24-6
C34H30N2O6S
594.68
AQXXZDYPVDOQEE-MXDQRGINSA-N
1-methyl-2-[(1E)-2-(thiophen-2-yl)ethenyl]-1,4,5,6-tetrahydropyrimidine; 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
CN1CCCN=C1\C=C\C1=CC=CS1.OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1
22204-24-6
C34H30N2O6S
594.68
AQXXZDYPVDOQEE-MXDQRGINSA-N
1-methyl-2-[(1E)-2-(thiophen-2-yl)ethenyl]-1,4,5,6-tetrahydropyrimidine; 4-[(3-carboxy-2-hydroxynaphthalen-1-yl)methyl]-3-hydroxynaphthalene-2-carboxylic acid
CN1CCCN=C1\C=C\C1=CC=CS1.OC(=O)C1=C(O)C(CC2=C3C=CC=CC3=CC(C(O)=O)=C2O)=C2C=CC=CC2=C1

P1599, 22204-24-6, C11H14N2S•C23H16O6

Spectrum™ Pyrantel Pamoate, USP is an anthelmintic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.005
Loss on Drying 0.02
Quantity 25 g
CAS Max % 1
Percent Purity 97 to 103%
Grade USP