Alfa Aesar™ Pyrazine-2-carbonitrile, 97+%

Manufacturer: Alfa Aesar™ A1097406

Quantity 5g

 View more versions of this product

Catalog No. AAA1097406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19847-12-2
    Molecular Formula C5H3N3
    Formula Weight 105.1
    MDL Number MFCD00049361
    UN Number UN3276
    Beilstein 110008
    Synonym 2-Cyanopyrazine
    Chemical Name or Material Pyrazine-2-carbonitrile

    Specifications

    Formula Weight 105.1
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 19847-12-2
    Synonym 2-Cyanopyrazine
    TSCA TSCA
    Molecular Formula C5H3N3
    MDL Number MFCD00049361
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Boiling Point 90° to 93°C (15mmHg)
    Melting Point 18° to 20°C
    Quantity 5g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
    Flash Point 96°C (204°F)
    UN Number UN3276
    Beilstein 110008
    Chemical Name or Material Pyrazine-2-carbonitrile
    Assay ≥97%

    Description

    Applications
    Pyrazine-2-carbonitrile is used as an organic chemical synthesis intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit