Alfa Aesar™ Pyrazine-2-thiocarboxamide, 97%

Manufacturer: Alfa Aesar™ H2675003

Quantity 1g

 View more versions of this product

Catalog No. AAH2675003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4604-72-2
    Molecular Formula C5H5N3S
    Formula Weight 139.17
    MDL Number MFCD00173663
    Synonym Pyrazine-2-carbothioamide
    Chemical Name or Material Pyrazine-2-thiocarboxamide

    Specifications

    Formula Weight 139.17
    MDL Number MFCD00173663
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Odor Odorless
    Melting Point 198° to 200°C
    Quantity 1g
    Molecular Formula C5H5N3S
    CAS 4604-72-2
    Synonym Pyrazine-2-carbothioamide
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material Pyrazine-2-thiocarboxamide
    Assay 97%

    Description

    Solubility
    Soluble in organic solvents, ethanol, DMSO.

    Notes
    Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit