Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyrene, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
129-00-0
C16H10
202.25
MFCD00004136
BBEAQIROQSPTKN-UHFFFAOYSA-N
benzo def phenanthrene,pyren,beta-pyrene,.beta.-pyrene,pyren german,unii-9e0t7wfw93,ccris 1256,pyrene def phenanthrene,coal tar pitch volatiles:pyrene
31423
CHEBI:39106
pyrene
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
129-00-0
C16H10
202.25
MFCD00004136
BBEAQIROQSPTKN-UHFFFAOYSA-N
benzo def phenanthrene,pyren,beta-pyrene,.beta.-pyrene,pyren german,unii-9e0t7wfw93,ccris 1256,pyrene def phenanthrene,coal tar pitch volatiles:pyrene
31423
CHEBI:39106
pyrene
C1=CC2=C3C(=C1)C=CC4=CC=CC(=C43)C=C2
Flash Point 210°C
Boiling Point 393°C
Melting Point 148°C to 152°C
Color Green-Yellow to Yellow
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Beilstein 05,693
Fieser 04,414
Merck Index 15,8074
Solubility Information Solubility in water: almost insoluble. Other solubilities: soluble in ethanol,ether,benzene and toluene,slightly soluble in carbon tetrachloride
Formula Weight 202.25
Physical Form Crystals or Chunks
Percent Purity 98%
Chemical Name or Material Pyrene
Show More Show Less

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.

GHS Signal Word: Warning

EINECSNumber : 204-927-3