Learn More
PYREX™ Accelerated One-Step™ Extractor/Concentrator Replacement Parts Available on GSA/VA Contract for Federal Government customers only.
Replacement parts for Accelerated One-Step Extractor/Concentrator Systems
Manufacturer:  Corning 3915MSCO
 View more versions of this product
Catalog No. 07-770-81
Add to cart
Description
Description
Replacement part for Pyrex Accelerated One-Step extractor/concentrator system. The system performs extraction, concentration, and drying with the same apparatus and is designed for use with solvents of heavier specific gravity than sample being extracted.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Snyder Sidearm
Pyrex™ accelerated One-Step™ extractor or concentrator system
24/40