Learn More
PYREX™ Biometer Culture Flask Available on GSA/VA Contract for Federal Government customers only.
Minimize the need for commonly used gas trains
Manufacturer:  Corning, Inc. 4443250
 View more versions of this product
Catalog No. 11-850-15
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Style: Biometer; Top style: Beaded rim
Delivery funnel, no.6 Rubber stopper in the flask neck, no.7 Rubber stopper in the side tube, 16-gauge needle
Non-sterile
Borosilicate Glass
Used to measure production of Carbon-di-oxide produced by microorganisms grown on a variety of culture medium
Wide
25mL