Learn More
Pyrex™ Brand Standard Wall Tubing
Low coefficient of expansion, great mechanical strength
Manufacturer:  Wale Apparatus CompanySupplier Diversity Partner 234050
 View more versions of this product
Catalog No. 11-365C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
U.S. pharmacopoeia
1219mm
0.8mm
2885°F
5mm
Pyrex™
0.19 in.
Standard Wall Tubing
±0.1mm
1252°C
48 in.