Learn More
Pyrex™ Brand Standard Wall Tubing
Low coefficient of expansion, great mechanical strength
Manufacturer:  Wale Apparatus CompanySupplier Diversity Partner 234250
 View more versions of this product
Catalog No. 11-365V
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
U.S. pharmacopoeia
1219mm
1.5mm
2885°F
25mm
Pyrex™
0.98 in.
Standard Wall Tubing
±0.2mm
1252°C
48 in.