Learn More
PYREX™ Buchner Funnels with Fritted Discs Available on GSA/VA Contract for Federal Government customers only.
Useful where filter paper would be attacked by material being filtered
Manufacturer:  Corning, Inc. 36060350C
 View more versions of this product
Catalog No. 10-358Q
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
11.83 oz.
ASTM E128
80mm
Fritted discs
11.83 oz., 350mL
20mm
180mm
75mm
40 to 60μm
350mL
3.14 in.
Useful where filter paper would be attacked by material being filtered
Coarse
0.78 in.
7.08 in.
2.95 in.
Pyrex™
Buchner Funnel