Learn More
PYREX™ Bumper Guards for Cylinders
Slide the ring toward the top of the cylinder to absorb shock, should the cylinder be accidentally upset.
Manufacturer: PYREX™ 3066100
Description
A polyethylene shock absorbing ring designed to reduce breakage of cylinders. For best results, slide the ring toward the top to absorb shock should the cylinder be accidentally upset.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Not Autoclavable | |
Fits cylinder capacity 100 mL, slide the ring toward the top to absorb shock should the cylinder be accidentally upset | |
Non-sterile |
100 mL | |
LDPE | |
Bumper Guard |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.