Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYREX™ Caps for Erlenmeyer Flasks Available on GSA/VA Contract for Federal Government customers only.

As supplied with Pyrex™ Brand Media/Solution bottles. Choose orange caps for temperatures to 140°C (284°F), red caps for temperatures to 180°C (356°F).

Manufacturer:  Corning 431363

 View more versions of this product

Catalog No. 07-200-865


Add to cart

Description

Description

  • Vented caps with 0.2um pore nonwettable (hydrophobic) membranes which allow gas exchange while minimizing risk of contamination
  • Polyethylene
  • Sterilized by gamma radiation

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Orange
Screw
7cm
3L Plastic Erlenmeyer flask
Sterile
140°C
Polypropylene
Style: Flat
Yes
Cap
284°F
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit