Learn More
PYREX™ Class B Mixing Cylinders Available on GSA/VA Contract for Federal Government customers only.
Calibrated “To Contain”
Manufacturer:  Corning 29822L
 View more versions of this product
Catalog No. 08-566H
Add to cart
Description
Description
  • Single metric scale
  • White-enamel graduations
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Borosilicate Glass
B
2000mL
23.22 in.
Single white enamel metric graduations
Barrelhead
Glass
Yes
ASTM
3.26 in.
8.3cm
590mm
38
±12mL
20mL
20mL