Learn More
PYREX™ Disposable Individually Wrapped Glass Serological Pipets Available on GSA/VA Contract for Federal Government customers only.
Calibrated “to deliver” (TD) with blow-out. Corning™ Pyrex™ Disposable Individually Wrapped Glass Serological Pipets offer long, slender, tapered tips to make pipetting go faster and easier.
Manufacturer:  Corning, Inc. 70771N
 View more versions of this product
Catalog No. 13-666-7F
  Check Availability
Add to cart
Specifications
Description
  • Sterile, plugged
  • Negative graduations
  • Fire-polished tips for burr-free, even flow rates
  • Easy-open Steri-View™ wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size
  • Colors conform to I.S.O. standards
Description & Specifications
Specifications
11.41 in.
0.2mL
Yes
Yellow
Serological Pipet
Round
290mm
1mL
glass
ASTM E714
Yes
0.01mL