Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PYREX™ Distilling Flasks Products for Science Education

Round bottoms, vial mouths, with side arms at 75° angle

$33.25 - $150.00

Specifications

Material Glass, Pyrex Glass
Length (Metric) Side Arm 13 cm
Neck Style Long
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity & Availability  
S34175 Products for Science Education
View Documents
PYREX™
4620250/EMD
250 mL
Pack of 4 for $150.00
Case of 6 PK for $899.64
Add to cart
 
S34176 Products for Science Education
View Documents
PYREX™
4620500/EMD
500 mL
Case for $33.25
Add to cart
 
Description

Description

The Pyrex™ name, synonymous with high-quality glassware, assures durability of these distilling flasks. Feature an extended arm.

Specifications

Specifications

Glass, Pyrex Glass
13 cm
Long
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit