Learn More
PYREX™ Economy Squibb Separatory Funnel, Polyethylene Standard Taper Stopper, Replaceable Standard PTFE Stopcock Available on GSA/VA Contract for Federal Government customers only.
Economical separatory funnel designed to reduce replacement costs by providing replaceable components
Manufacturer:  Corning, Inc. 6406125
 View more versions of this product
Catalog No. 10-444-11
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
4.22 oz.
6.03cm
Economy Squibb Separatory Funnel
Polyethylene
125mL
0.27 in.
2.04 in.
Pyrex™
2.37 in.
For standard taper stoppers and for PTFE locking stopcock assemblies
4 Case
22
4.22 oz., 125mL
0.7cm
52mm
4