PYREX™ Enlarging Glass Joint Adapters Available on GSA/VA Contract for Federal Government customers only.
For attaching apparatus with one standard taper joint size to equipment having larger size standard taper joint
Manufacturer: Corning, Inc. 8820-1019
 View all options for this product
Catalog No. 01-034A
  Check Availability
Add to Cart
    Specifications
    Product Type Tube Adapter
    Material Pyrex™
    Standard Taper Joint No. 19/38 Inner Bottom, 10/30 Outer Top
    For Use With (Equipment) For attaching apparatus with one joint size to equipment having larger size joint
    Length (English) 3.34 in.
    Length (Metric) 85mm
    • Outer standard taper joint at top and larger inner standard taper joint at bottom
  • Description & Specifications
Related Products
Product Content Feedback