Learn More
PYREX™ Filter Tubes Available on GSA/VA Contract for Federal Government customers only.
Used to collect filtrate
Manufacturer:  Corning 948042
 View more versions of this product
Catalog No. 08-261D
Add to cart
Description
Description
  • Use on the bottom end of 30mm (1.18 in.) O.D. Pyrex Gooch Low Form fritted disc crucibles (Cat. Nos. 08-236-1A, B, C)
  • Also for use with porcelain plates
Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1.73 inch
42mm
Used to collect the filtrate
6.29 in.
Filter Thimble
0.31 in.
0.8cm
Pyrex gooch fritted discs crucibles and porcelain plates
16cm
Pyrex