PYREX™ Flash Chromatography Column, Spherical Joint Socket Joint, PTFE stopcock and 0-4mm PTFE valve Available on GSA/VA Contract for Federal Government customers only.
For quick separations in absorption chromatography applications
Manufacturer: Corning, Inc.Supplier Diversity Partner 214922
 View more versions of this product
Catalog No. 05-723-25B
  Check Availability
Add to Cart
    Specifications
    Column Type Flash chromatography Column
    Construction 35/20 Spherical Joint socket joint at the top and with a 0 to 4mm PTFE plug stopcock at the bottom
    Diameter (English) Outer 0.86 in.
    Diameter (Metric) Outer 22mm
    For Use With (Equipment) For flash reservoir and a flow controller
    Length (Metric) 450mm
    Material Pyrex Glass
    Style Straight Bore PTFE
  • Description & Specifications
Related Products
Product Content Feedback