Learn More
PYREX™ Flash Chromatography Column, Spherical Joint Socket Joint, PTFE stopcock and 0-4mm PTFE valve
For quick separations in absorption chromatography applications
Manufacturer:  Corning 214954
 View more versions of this product
Catalog No. 05-723-25D
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 35/20 Spherical Joint socket joint at top and with 0-4mm PTFE plug stopcock at bottom
  • Designed to be used with Flash reservoir (0572324 series) and flow controller
Specifications
Specifications
450 mm
Straight Bore PTFE
54 mm
Flash Chromatography Column
Pyrex Glass
2.12 in.
35/20 Spherical Joint socket joint at the top and with a 0 to 4 mm PTFE plug stopcock (at the bottom)
For Flash Reservoir and a Flow Controller