Learn More
PYREX™ Flasks with standard taper Polyethylene Stoppers Available on GSA/VA Contract for Federal Government customers only.
Sharp, clear graduation lines
Manufacturer:  Corning, Inc. 5642250
 View more versions of this product
Catalog No. 10-210-5E
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Flat
16
78mm
25.2cm
±0.12mL
Glass,Pyrex™
HDPE
3.07 in.
9.92 in.
Long
250mL