Learn More
PYREX™ Glass Extraction Thimbles Available on GSA/VA Contract for Federal Government customers only.
Resist attack by most chemicals
Manufacturer:  Corning 33950MC
 View more versions of this product
Catalog No. 09-653B
Add to cart
Description
Description
  • Borosilicate glass
  • With fritted disc of coarse (nominal max. pore size, 40-60µm) or extra-coarse (nominal max. pore size, 170-220µm) porosity
  • Fit into standard Soxhlet extraction tubes
  • Convenient for weighing extracted materials
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1.37 in.
For weighing extracted materials
3.54 in.
Fritted discs
Coarse
Borosilicate Glass
3.5cm
For Pyrex extractors and extraction apparatus, soxhlet extractors
9cm
40 to 60μm
Extraction Thimble